Anti-MSR1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301566EA 569 EUR
ORIGTA301566
Anti-MSR1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: MSR1 (macrophage scavenger receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR