Anti-DNMT3A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301633EA 624 EUR
ORIGTA301633
Anti-DNMT3A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: DNMT3A (DNA (cytosine-5-)-methyltransferase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR