Anti-KIAA1524 Mouse Monoclonal Antibody [clone: 2G10]

Supplier: OriGene
ORIGTA301640EA 545 EUR
ORIGTA301640
Anti-KIAA1524 Mouse Monoclonal Antibody [clone: 2G10]
Antibodies
Recommended Dilutions: Western Blot: 0.5 ?g/ml; Immunofluorescence: 1: 100

Type: Primary
Antigen: KIAA1524 (KIAA1524)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR