Anti-TPX2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301667EA 545 EUR
ORIGTA301667
Anti-TPX2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: TPX2 (TPX2, microtubule-associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR