Anti-IMMT Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301674EA 747 EUR
ORIGTA301674
Anti-IMMT Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: IMMT (inner membrane protein, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR