Anti-TERF2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301691EA 747 EUR
ORIGTA301691
Anti-TERF2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:2000-1:5000

Type: Primary
Antigen: TERF2 (telomeric repeat binding factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR