Anti-INSIG1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301693EA 650 EUR
ORIGTA301693
Anti-INSIG1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: INSIG1 (insulin induced gene 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR