Anti-CTGF Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301728EA 658 EUR
ORIGTA301728
Anti-CTGF Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1: 1000-1:2000

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR