Anti-BUB1B Mouse Monoclonal Antibody [clone: 8G1]

Supplier: OriGene
ORIGTA301741EA 601 EUR
ORIGTA301741
Anti-BUB1B Mouse Monoclonal Antibody [clone: 8G1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000-1:5000

Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR