Anti-ATP7B Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301761EA 590 EUR
ORIGTA301761
Anti-ATP7B Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: ATP7B (ATPase, Cu++ transporting, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR