Anti-MRE11A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301767EA 658 EUR
ORIGTA301767
Anti-MRE11A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000; Immunofluorescence: 1:200

Type: Primary
Antigen: MRE11A (MRE11 meiotic recombination 11 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR