Anti-LOX Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301771EA 447 EUR
ORIGTA301771
Anti-LOX Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000-1:5000; Immunochemistry: 10-20 ?g/ml

Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR