Anti-BIRC5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301775EA 650 EUR
ORIGTA301775
Anti-BIRC5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR