Anti-HSPA1A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302005EA 698 EUR
ORIGTA302005
Anti-HSPA1A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HSPA1A (heat shock 70kDa protein 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR