Anti-EIF4EBP1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302037EA 579 EUR
ORIGTA302037
Anti-EIF4EBP1 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR