Anti-PDHX Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302073EA 638 EUR
ORIGTA302073
Anti-PDHX Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100; Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PDHX (pyruvate dehydrogenase complex, component X)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR