Anti-NR0B1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302083EA 698 EUR
ORIGTA302083
Anti-NR0B1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NR0B1 (nuclear receptor subfamily 0, group B, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR