Anti-GNB2L1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302104EA 638 EUR
ORIGTA302104
Anti-GNB2L1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: GNB2L1 (guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR