Anti-UBQLN1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302118EA 638 EUR
ORIGTA302118
Anti-UBQLN1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: UBQLN1 (ubiquilin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR