Anti-RAC1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302136EA 579 EUR
ORIGTA302136
Anti-RAC1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RAC1 (ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR