Anti-ATOH1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302151EA 579 EUR
ORIGTA302151
Anti-ATOH1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ATOH1 (atonal homolog 1 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR