Anti-RPS6KB1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302199EA 579 EUR
ORIGTA302199
Anti-RPS6KB1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: RPS6KB1 (ribosomal protein S6 kinase, 70kDa, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR