Anti-AKT2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302205EA 579 EUR
ORIGTA302205
Anti-AKT2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR