Anti-MAP2K1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302208EA 698 EUR
ORIGTA302208
Anti-MAP2K1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR