Anti-MTOR Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302216EA 579 EUR
ORIGTA302216
Anti-MTOR Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: MTOR (mechanistic target of rapamycin (serine/threonine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR