Anti-CASK Rabbit polyclonal antibody

Supplier: OriGene
ORIGTA302217EA 698 EUR
ORIGTA302217
Anti-CASK Rabbit polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: CASK (calcium/calmodulin-dependent serine protein kinase (MAGUK family))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR