Anti-ATF4 Rabbit polyclonal antibody

Supplier: OriGene
ORIGTA302232EA 698 EUR
ORIGTA302232
Anti-ATF4 Rabbit polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ATF4 (activating transcription factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR