Anti-TAOK3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302258EA 698 EUR
ORIGTA302258
Anti-TAOK3 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: TAOK3 (TAO kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR