Anti-PHPT1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302277EA 638 EUR
ORIGTA302277
Anti-PHPT1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PHPT1 (phosphohistidine phosphatase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR