Anti-CNOT4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302282EA 698 EUR
ORIGTA302282
Anti-CNOT4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CNOT4 (CCR4-NOT transcription complex, subunit 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR