Anti-ZIC4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302374EA 579 EUR
ORIGTA302374
Anti-ZIC4 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ZIC4 (Zic family member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR