Anti-ACADM Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302400EA 454 EUR
ORIGTA302400
Anti-ACADM Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 001-01µg/ml

Type: Primary
Antigen: ACADM (acyl-CoA dehydrogenase, C-4 to C-12 straight chain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR