Anti-ALDH1A1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302409EA 454 EUR
ORIGTA302409
Anti-ALDH1A1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: ALDH1A1 (aldehyde dehydrogenase 1 family, member A1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR