Anti-COPS2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302410EA 547 EUR
ORIGTA302410
Anti-COPS2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1µg/ml

Type: Primary
Antigen: COPS2 (COP9 signalosome subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR