Anti-APBB1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302412EA 454 EUR
ORIGTA302412
Anti-APBB1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: APBB1 (amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR