Anti-MANF Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302417EA 500 EUR
ORIGTA302417
Anti-MANF Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: MANF (mesencephalic astrocyte-derived neurotrophic factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR