Anti-ASF1A Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302423EA 547 EUR
ORIGTA302423
Anti-ASF1A Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:1000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: ASF1A (anti-silencing function 1A histone chaperone)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR