Anti-BGN Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302434EA 454 EUR
ORIGTA302434
Anti-BGN Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: BGN (biglycan)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR