Anti-CSNK1D Goat polyclonal antibody

Supplier: OriGene
ORIGTA302441EA 454 EUR
ORIGTA302441
Anti-CSNK1D Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: CSNK1D (casein kinase 1, delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR