Anti-CHRNA4 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302451EA 454 EUR
ORIGTA302451
Anti-CHRNA4 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: CHRNA4 (cholinergic receptor, nicotinic, alpha 4 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR