Anti-DAP3 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302460EA 454 EUR
ORIGTA302460
Anti-DAP3 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3 ?g/ml; Immunochemistry: 3-10?g/ml

Type: Primary
Antigen: DAP3 (death associated protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR