Anti-DAPK2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302461EA 500 EUR
ORIGTA302461
Anti-DAPK2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: DAPK2 (death-associated protein kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR