Anti-DDC Goat polyclonal antibody

Supplier: OriGene
ORIGTA302469EA 550 EUR
ORIGTA302469
Anti-DDC Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: DDC (dopa decarboxylase (aromatic L-amino acid decarboxylase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR