Anti-NR1H4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302484EA 454 EUR
ORIGTA302484
Anti-NR1H4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: NR1H4 (nuclear receptor subfamily 1, group H, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR