Anti-FGR Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302485EA 454 EUR
ORIGTA302485
Anti-FGR Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-2µg/ml

Type: Primary
Antigen: FGR (feline Gardner-Rasheed sarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR