Anti-FLOT1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302489EA 547 EUR
ORIGTA302489
Anti-FLOT1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 3-5?g/ml; ELISA: 1:32000

Type: Primary
Antigen: FLOT1 (flotillin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR