Anti-GCNT3 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302497EA 547 EUR
ORIGTA302497
Anti-GCNT3 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: GCNT3 (glucosaminyl (N-acetyl) transferase 3, mucin type)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR