Anti-GPR119 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302500EA 456 EUR
ORIGTA302500
Anti-GPR119 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: GPR119 (G protein-coupled receptor 119)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR