Anti-GNRHR Goat polyclonal antibody

Supplier: OriGene
ORIGTA302503EA 500 EUR
ORIGTA302503
Anti-GNRHR Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: GNRHR (gonadotropin-releasing hormone receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR