Anti-GSTP1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302504EA 454 EUR
ORIGTA302504
Anti-GSTP1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: GSTP1 (glutathione S-transferase pi 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR